Orders LinkContactUsLinkComponent
Small flanges with hose nozzle
Small flanges with hose nozzle
06, Small flanges, DN 25, Hliník, 10 mm
  185-2254
 :   VACUUBRAND
Small flanges with hose nozzle
  185-2254
Small flanges with hose nozzle
Small flanges with hose nozzle
06, Small flanges, DN 25, Hliník, 10 mm
838,00 Kč
1 * 1 KS
These flanges are made from aluminium or plastic.
  • Fast, easy assembly and disassembly to/from pumps
  • Safe, vacuum tight connection with optional centring and clamping rings
  • A wide range of flanges with hose nozzles for various internal hose diameters are available
For connecting pumps to vacuum hoses.
  • Hadice int.Ø:
    10 mm
  • Popis:
    Small flanges, DN 25
  • Materiál:
    Hliník
  • Sekvence:
    06