Orders LinkContactUsLinkComponent
Filter pump
Filter pump
Water pump 5701, with non return valve (165 g)
Katalogové číslo 181-0117
Dodavatel:   USBECK Laborgeräte
Filter pump
Katalogové číslo 181-0117
Dodavatel:  USBECK Laborgeräte
Filter pump
Filter pump
Water pump 5701, with non return valve (165 g)
Price
1 890,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Water jet pump in nickel-plated brass.
  • Stable and solid
  • A side knob allows connection to a vacuum pipe with an interior diameter of 8 mm
  • Available with or without non-return valve
  • Supplied with water connection for pipes with an internal diameter of 14 mm or bolting for connection to a tap
  • Pressure gauge available as an option for all models
Documents
Specifikace
  • Popis:
    Water pump 5701, with non return valve (165 g)
Accessories
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Water pump manometer
    Water pump manometer
    181-0121
    Přihlásit se pro dostupnost
    1 * 1 KS - 2 000,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • G 1/2 connection for water pump
    G 1/2 connection for water pump
    USBE5741
    Přihlásit se pro dostupnost
    1 * 1 KS - 675,00 Kč