Hoffmann pinch cocks
20 mm
Katalogové číslo 229-0343
Dodavatel: USBECK Laborgeräte
Přejít na:
Product Details & Documents
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
- Hinged
- Very good thermal resistance
- The standard model has a knurled screw and the heavy duty model has a butterfly screw
- Easy cleaning
Informace pro objednávání: The values in the band width column refer to the clearance between the pinch cock legs.
Specifikace
- Šířka proužku:20 mm