Orders LinkContactUsLinkComponent
Hoffmann pinch cocks
Hoffmann pinch cocks
12 mm
Katalogové číslo 229-0341
Dodavatel:   USBECK Laborgeräte
Hoffmann pinch cocks
Katalogové číslo 229-0341
Dodavatel:  USBECK Laborgeräte
Hoffmann pinch cocks
Hoffmann pinch cocks
12 mm
Přejít na:
Price
164,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Hinged
  • Very good thermal resistance
  • The standard model has a knurled screw and the heavy duty model has a butterfly screw
  • Easy cleaning
Ordering information: The values in the band width column refer to the clearance between the pinch cock legs.
Specifikace
  • Šířka proužku:
    12 mm