Orders LinkContactUsLinkComponent
Tripod stands
Tripod stands
11, 140×220 mm
Catalog # 451-0094
Supplier:   USBECK Laborgeräte
Tripod stands
Catalog # 451-0094
Tripod stands
Tripod stands
11, 140×220 mm
Price
492,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.
Specifications
  • Vnitř. Ø×V:
    140×220 mm
  • Sekvence:
    11