Blunt forceps
Tweezers, 130 mm
232-0104
Stainless steel.
- Curved
- Blunt pointed ends
- Popis:Tweezers
- Délka:130 mm
Kyselina 1-pentansulfanová sodná sůl ≥99%, HiPerSolv CHROMANORM® for HPLC
#: 152813W
: VWR Chemicals
broman draselný ≥99.8% (dry basis), AnalaR NORMAPUR® analytical reagent
#: 26703.232
: VWR Chemicals
Dihydrogenfosforečnan sodný dihydrát ≥99.0%, AnalaR NORMAPUR® analytical reagent
#: 28015.294
: VWR Chemicals