Bosshead
Hinged screws, 13 mm, Slitina hliníku, práškované
Catalog # 241-7231
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
For rods up to 13 mm. Adjustable, nickel plated brass screws.
Specifications
- Upínací šířka:13 mm
- Popis:Hinged screws
- Materiál:Slitina hliníku, práškované