Stand plates
300×150 mm, 2,8 kg
Katalogové číslo 241-7112
Dodavatel: USBECK Laborgeräte
Přejít na:
Product Details & Documents
Steel, powder coated. Firm, non slip stand. For a wide variety of uses. With M10 threaded hole on face side.
- Rubber feet for a firm footing
- Easy to clean
- Robust
Specifikace
- D׊:300×150 mm
- Hmotnost:2,8 kg