Micro burners with needle valve
Micro burner with needle valve, Natural gas, 11 mm, 1450 °C
Katalogové číslo 451-0084
Dodavatel: USBECK Laborgeräte
Přejít na:
Product Details & Documents
Micro burner with air regulation and needle valve.
Certifications: DIN 30665
Specifikace
- Max. tepl.:1450 °C
- Popis:Micro burner with needle valve
- Ø hlavy:11 mm
- Určeno pro:Natural gas