Orders LinkContactUsLinkComponent
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, sterile, Permanox™, 27×75 mm
Catalog # 734-2071
   Low Manufacturing Impact
Microscope slides, Nunc™
Catalog # 734-2071
   Low Manufacturing Impact
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, sterile, Permanox™, 27×75 mm
Price
13 450,00 Kč
Log in for availability
1 * 100 KS
Product Details & Documents
These microscope slides are designed for applications in cell culture.
The surface treatment ensures optimal conditions for cell attachment and growth.
Specifications
  • Popis:
    Microscope slides, sterile, Permanox™
  • D׊:
    27×75 mm
  • Cat. No.:
    734-2071
  • Certifikát ochrany životního prostředí:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact