Orders LinkContactUsLinkComponent
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, sterile, PS, 25×75 mm
Katalogové číslo 734-2070
   Low Manufacturing Impact
Microscope slides, Nunc™
Katalogové číslo 734-2070
   Low Manufacturing Impact
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, sterile, PS, 25×75 mm
Přejít na:
Price
16 150,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
These microscope slides are designed for applications in cell culture.
The surface treatment ensures optimal conditions for cell attachment and growth.
Specifikace
  • Popis:
    Microscope slides, sterile, PS
  • D׊:
    25×75 mm
  • Cat. No.:
    734-2070
  • Certifikát ochrany životního prostředí:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact