Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Test kits, ammonium, Spectroquant®, Supelco®
Test kits, ammonium, Spectroquant®, Supelco®
03, Amoniový kyvetový test, 0,5−16,0 mg/L NH₄ – N, 0,6−20,6 mg/L NH₄, Indofenolová modr, Prove, Nova 30/60, 25 Tests
Catalog # 1.14544.0001
Supplier:   Merck
Test kits, ammonium, Spectroquant®, Supelco®
Catalog # 1.14544.0001
Supplier:  Merck
CAS Number:  
Test kits, ammonium, Spectroquant®, Supelco®
Test kits, ammonium, Spectroquant®, Supelco®
03, Amoniový kyvetový test, 0,5−16,0 mg/L NH₄ – N, 0,6−20,6 mg/L NH₄, Indofenolová modr, Prove, Nova 30/60, 25 Tests
CAS Number:  
Price
4 360,00 Kč
Log in for availability
1 * 25 Tests
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
ISO 23695: Water Quality — Determination of Ammonium Nitrogen in Water — Small-Scale Sealed Tube Method
• Method: This standard utilizes the Bertholet method (Indophenol blue) for determining ammonium.
• Citation Forms: Various forms such as NH₄⁺, NH₄-N, NH₃, NH₃-N are acceptable.
• Spectroquant® Cell Tests 1.14544, 1.14558, 1.14559, and 1.14739 correspond to this norm using 2-Chlorphenol.
Specifications
Specifications
  • Chemická metoda:
    Indofenolová modr
  • Bal.:
    25 Tests
  • Popis:
    Amoniový kyvetový test
  • Sekvence:
    03
  • Pro jednotku:
    Prove, Nova 30/60
  • Rozsah:
    0,5−16,0 mg/L NH₄ – N, 0,6−20,6 mg/L NH₄
  • Cat. No.:
    1.14544.0001
  • M-Clarity quality level:
    MQ100
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.