Orders LinkContactUsLinkComponent
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
25, Kyvetový test COD*⁷⁻¹, 25−1 500 mg/L, Chromosulfurická oxidace, Prove, Nova 30/60, Multy, Move, Picco, 25 Tests
Katalogové číslo 1.14541.0001
Dodavatel:   Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Katalogové číslo 1.14541.0001
Dodavatel:  Merck
Číslo CAS:  
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
25, Kyvetový test COD*⁷⁻¹, 25−1 500 mg/L, Chromosulfurická oxidace, Prove, Nova 30/60, Multy, Move, Picco, 25 Tests
Číslo CAS:  
Price
4 630,00 Kč
Přihlásit se pro dostupnost
1 * 25 Tests
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and wastewater
*8 COD cell tests fully comply with ISO 15705.
Specifikace
Specifikace
  • Chemická metoda:
    Chromosulfurická oxidace
  • Bal.:
    25 Tests
  • Popis:
    Kyvetový test COD*⁷⁻¹
  • Sekvence:
    25
  • Pro jednotku:
    Prove, Nova 30/60, Multy, Move, Picco
  • Rozsah:
    25−1 500 mg/L
  • Cat. No.:
    1.14541.0001
  • M-Clarity quality level:
    MQ100
Specifikace
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Bal.
    Dostupnost
    Price
  • Nástavce dávkovací k CHSK analýze mořské vody 1.17059.0001
    Nástavce dávkovací k CHSK analýze mořské vody 1.17059.0001
    1.15955.0001
    02
    1
    Přihlásit se pro dostupnost
    1 * 1 KS - 6 790,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Typ balení
    Bal.
    Dostupnost
    Price
  • 1.06733.0501
    3645
    Plastová láhev
    500 g
    Přihlásit se pro dostupnost
    1 * 500 g - 2 780,00 Kč