Orders LinkContactUsLinkComponent
Modular Storage Drawers
Modular Storage Drawers
01, Drawers only, 40,5 × 23 × 5,1 cm
Katalogové číslo 720-0280
Dodavatel:   Simport Scientific
Modular Storage Drawers
Katalogové číslo 720-0280
Dodavatel:  Simport Scientific
Modular Storage Drawers
Modular Storage Drawers
01, Drawers only, 40,5 × 23 × 5,1 cm
Přejít na:
Price
3 540,00 Kč
Přihlásit se pro dostupnost
1 * 6 KS
Product Details & Documents
Storage drawers from PS. Permanent storage of up to 165 embedding rings or 250 cassettes per drawer.
  • Interlocking ridges for stacking
  • Identification labels included
Specifikace
  • Popis:
    Drawers only, 40,5 × 23 × 5,1 cm
  • Sekvence:
    01