pH indicator strips, MQuant®, Supelco®
08, Special pH indicator strips, 6,5 - 10,0, 0,2 - 0,5
Dodavatel: Merck
Přejít na:
Product Details & Documents
These test strips contain special indicator dyes that are covalently bound to the reagent papers which are then adhered to the plastic strips.
- Non bleeding - do not contaminate the solution to be measured
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
: Analyse your pH 0 to 14 test strips (1.09535) with this reference card and the free MQuant® StripScan app. The brilliant colour-fast reference colours allow for robust measurement results even under changing lighting conditions. The StripScan app allows the customer to analyse the MQuant® test strips with their smartphone for fast, convenient and precise results which can be easily graphically presented and shared.
Specifikace
- pH stupnice:0,2 - 0,5
- pH rozsah:6,5 - 10,0
- Popis:Special pH indicator strips
- Sekvence:08
- Cat. No.:1.09543.0001
- M-Clarity quality level:MQ100