pH indicator papers, MQuant®, Supelco®
Universal indicator replacement rolls only*, 1 - 10, 1
Dodavatel: Merck
Přejít na:
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
Documents
Specifikace
- pH stupnice:1
- pH rozsah:1 - 10
- Popis:Universal indicator replacement rolls only*
- Cat. No.:1.09527.0001
- M-Clarity quality level:MQ100