Orders LinkContactUsLinkComponent
Base materials
21, Peptone from meat, pancreatic, granulated, 1 kg
Katalogové číslo 1.07214.1000
Dodavatel:   Merck
Base materials
Katalogové číslo 1.07214.1000
Dodavatel:  Merck
Číslo CAS:  
Base materials
21, Peptone from meat, pancreatic, granulated, 1 kg
Přejít na:
Číslo CAS:  
Price
12 990,00 Kč
Přihlásit se pro dostupnost
1 * 1 kg
Product Details & Documents
Merck products offer high quality for applications in the microbiology field.
  • All products from one manufacturer
  • Constant quality and low costs as a result of bulk buying
  • Custom manufacture following customer developed protocols with excellent quality control
According DIN ISO 9001-2000 and ISO 11133.
 : According DIN ISO 9001-2000 and ISO 11133.
Specifikace
Specifikace
  • Bal.:
    1 kg
  • Popis:
    Peptone from meat, pancreatic, granulated
  • Sekvence:
    21
Specifikace