Orders LinkContactUsLinkComponent
Base materials
Base materials
05, Gelatin, 500 g
Katalogové číslo 1.04070.0500
Dodavatel:   Merck
Base materials
Katalogové číslo 1.04070.0500
Dodavatel:  Merck
Číslo CAS:  
Base materials
05, Gelatin, 500 g
Přejít na:
Číslo CAS:  
Price
3 890,00 Kč
Přihlásit se pro dostupnost
1 * 500 g
Product Details & Documents
Merck products offer high quality for applications in the microbiology field.
  • All products from one manufacturer
  • Constant quality and low costs as a result of bulk buying
  • Custom manufacture following customer developed protocols with excellent quality control
According DIN ISO 9001-2000 and ISO 11133.
Certifications: According DIN ISO 9001-2000 and ISO 11133.
Specifikace
Specifikace
  • Bal.:
    500 g
  • Popis:
    Gelatin
  • Sekvence:
    05