Jars, multipurpose, with covers, Nalgene®
8300 ml, 222 mm, 258 mm
Katalogové číslo 216-8265
Dodavatel: Thermo Fisher Scientific
Jars, multipurpose, with covers, Nalgene®
Katalogové číslo 216-8265
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
Polycarbonate, clear.
- Drill, tap or apply clamps to jars to attach heating or cooling equipment
- Very durable
- Autoclavable
Multipurpose jars with cover, which can be used as water baths, aquaria/terraria or freeze-drying chambers.
Documents
Specifikace
- Ø vněj.:222 mm
- Kapacita:8300 ml
- Výška:258 mm
- Cat. No.:216-8265
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact