Adapter, stopper type, DURAN®
zátka, dutá, 29/32 NS, 19/26 NS, ,
Katalogové číslo SCOT286576803
Dodavatel: DWK Life Sciences
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.
Specifikace
- Rozměry zábrusu:29/32 NS
- Velikost objímky:19/26 NS
- Type:zátka, dutá