Orders LinkContactUsLinkComponent
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
zátka, pevná, 29/32 NS, 14/23 NS, ,
Catalog # SCOT286576503
Supplier:   DWK Life Sciences
Adapter, stopper type, DURAN®
Catalog # SCOT286576503
Supplier:  DWK Life Sciences
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
zátka, pevná, 29/32 NS, 14/23 NS, ,
Price
10 750,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.
Specifications
  • Rozměry zábrusu:
    29/32 NS
  • Velikost objímky:
    14/23 NS
  • Type:
    zátka, pevná