Orders LinkContactUsLinkComponent
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
49, Cover glasses, 26×76 mm, #1,5
Katalogové číslo MENZBC026076AC40
Dodavatel:   Epredia
Rectangular cover glasses, Menzel Gläser
Katalogové číslo MENZBC026076AC40
Dodavatel:  Epredia
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
49, Cover glasses, 26×76 mm, #1,5
Přejít na:
Price
11 410,00 Kč
Přihlásit se pro dostupnost
1 * 1.000 KS
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles or flaws
  • Made of pre-washed pure white Hydrolytic Class 1 glass
Specifikace
  • Počet:
    #1,5
  • Popis:
    Cover glasses
  • D׊:
    26×76 mm
  • Sekvence:
    49