Orders LinkContactUsLinkComponent
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
26, Cover glasses, 24×50 mm, #2
Catalog # 630-2867
Supplier:   Epredia
Rectangular cover glasses, Menzel Gläser
Catalog # 630-2867
Supplier:  Epredia
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
26, Cover glasses, 24×50 mm, #2
Price
2 910,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles or flaws
  • Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
Specifications
  • Počet:
    #2
  • Popis:
    Cover glasses
  • D׊:
    24×50 mm
  • Sekvence:
    26