Orders LinkContactUsLinkComponent
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
04, Cover glasses, 21×26 mm, #1
Catalog # 631-1335
Supplier:   Epredia
Rectangular cover glasses, Menzel Gläser
Catalog # 631-1335
Supplier:  Epredia
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
04, Cover glasses, 21×26 mm, #1
Price
1 320,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles or flaws
  • Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
Specifications
  • Počet:
    #1
  • Popis:
    Cover glasses
  • D׊:
    21×26 mm
  • Sekvence:
    04