Nalgene™ Screw Caps
46, Replacement closure, 53 B, Jantarová, PP
Katalogové číslo NALG71-2171-0530
Dodavatel: Thermo Fisher Scientific
Nalgene™ Screw Caps
Katalogové číslo NALG71-2171-0530
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
PP, coloured.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifikace
- Velikost víčka:53 B
- Barva:Jantarová
- Popis:Replacement closure
- Materiál:PP
- Sekvence:46
- Cat. No.:NALG71-2171-0530
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact