Orders LinkContactUsLinkComponent
Cables for Electrodes
Cables for Electrodes
24, Plug cable combination, LB1A (coaxial electrode plug (for BlueLine), 1 m coaxial cable, DIN instrument plug), 1 m
Katalogové číslo 662-6144
Dodavatel:   SI Analytics
Cables for Electrodes
Katalogové číslo 662-6144
Dodavatel:  SI Analytics
Cables for Electrodes
Cables for Electrodes
24, Plug cable combination, LB1A (coaxial electrode plug (for BlueLine), 1 m coaxial cable, DIN instrument plug), 1 m
Přejít na:
Price
1 980,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Specifikace
  • Popis:
    Plug cable combination, LB1A (coaxial electrode plug (for BlueLine), 1 m coaxial cable, DIN instrument plug)
  • Délka:
    1 m
  • Sekvence:
    24