Reference electrodes
Reference electrode, low maintenance, Ag/AgCl reference system, B 1881 HD, −5...+100 °C, PVDF, Paste, PVDF, 12×120 mm, Screw plug head
Katalogové číslo SCOI285111767
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Produktová literatura
Specifikace
- Připojení:Screw plug head
- Ø×D:12×120 mm
- Type:B 1881 HD
- Popis:Reference electrode, low maintenance, Ag/AgCl reference system
- Materiál:PVDF
- Diafragma:PVDF
- Elektrolyt:Paste
- Tepl.:−5...+100 °C