Process pH combination electrodes
09, pH combination electrode, with sphere membrane, H 9080, 1 - 14, 0...+135 °C, Sklo, KCl 3 mol/l, Ceramic, 12×120 mm, Screw plug head
Katalogové číslo SCOI285110795
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product Literature
Specifikace
- Připojení:Screw plug head
- Ø×D:12×120 mm
- Type:H 9080
- Popis:pH combination electrode, with sphere membrane
- Materiál:Sklo
- Sekvence:09
- Diafragma:Ceramic
- Elektrolyt:KCl 3 mol/l
- Tepl.:0...+135 °C
- pH:1 - 14