Separating funnels, Squibb, Nalgene®
250 ml, 33 mm
Katalogové číslo 527-0006
Dodavatel: Thermo Fisher Scientific
Separating funnels, Squibb, Nalgene®
Katalogové číslo 527-0006
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
PPCO translucent, TFE stopcock, PP screw closure.
- Leakproof closure and stopcock
- Very good chemical resistance
- Autoclavable except for stopcock assembly
- TFE stopcock requires no lubrication
Specifikace
- Kapacita:250 ml
- Ø víčka:33 mm
- Cat. No.:527-0006
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact