Büchner funnels, Nalgene®
2906 ml, 150 mm
Catalog # 221-0104
Supplier: Thermo Fisher Scientific
Büchner funnels, Nalgene®
Catalog # 221-0104
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP
- Robust and lightweight
- Top and bottom can be separated for easy cleaning
- Autoclavable
For filter paper or membrane filtration discs.
Specifications
- Kapacita:2906 ml
- Pro filtr Ø:150 mm
- Cat. No.:221-0104
- Certifikát ochrany životního prostředí:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact