Volumetric flasks, class B, Nalgene®
100 ml, 0,16 ± ml
Katalogové číslo 612-3022
Dodavatel: Thermo Fisher Scientific
Volumetric flasks, class B, Nalgene®
Katalogové číslo 612-3022
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
PMP (TPX®), transparent, with PP screw cap.
- Very robust and allowing precise measurement
- Calibrated individually with permanent colour graduation line
- Autoclavable when the stopper has been removed from the threaded neck and a small quantity of water has been added to the flask. Cleaning with mild alkaline detergents recommended up to +60 °C (accuracy may be affected over +60 °C)
ASTM E288 and ISO Standard 384 Class B
Certifications: ASTM E288 and ISO Standard 384 Class B
Specifikace
- Kapacita:100 ml
- Tolerance:0,16 ± ml
- Cat. No.:612-3022
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact