Orders LinkContactUsLinkComponent
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
100 ml, 19/26 NS
Katalogové číslo SCOT242942409
Dodavatel:   DWK Life Sciences
Conical separating funnels, DURAN®
Katalogové číslo SCOT242942409
Dodavatel:  DWK Life Sciences
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
100 ml, 19/26 NS
Přejít na:
Price
2 210,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with NS-PE stopper.
  • Very good chemical resistance
  • Very good temperature resistance
  • Wwithout graduation complies to DIN ISO 4800
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance.
Specifikace
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    100 ml