Büchner funnels, DURAN®
73 mm, 220 ml, 70 mm
Catalog # 511-0337
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
For filter papers or membrane filtration discs.
Specifications
- Ø destičky:73 mm
- Kapacita:220 ml
- Pro filtr Ø:70 mm