Swabs
Non sterile swabs, Plastové/bavlna, 2,2 mm, 150 mm
Catalog # LPIT112491
Supplier: LP ITALIANA
Jump to:
Product Details & Documents
Sterile and non sterile swabs designed for use in microbiology.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Specifications
- Ø:2,2 mm
- Popis:Non sterile swabs
- Délka:150 mm
- Shaft/head:Plastové/bavlna