Vacuum filter holders
01, 85 ml, 47 mm
Catalog # SART16522
Supplier: Sartorius
Jump to:
Product Details & Documents
These vacuum filter holders feature a glass cylinder, PP base and sealing plug with anodised aluminum closing cap.
- Vacuum operated
Specifications
- Pro Ø:47 mm
- Kapacita:85 ml
- Sekvence:01
Accessories
Product Information
- Catalog #PopisAvailabilityPrice
Product Information
- Catalog #PopisAvailabilityPrice
Product Information
- Catalog #PopisAvailabilityPrice
Product Information
- Catalog #PopisAvailabilityPrice
Product Information
- Catalog #PopisAvailabilityPrice
Product Information
- Catalog #PopisAvailabilityPrice
Product Information
- Catalog #PopisAvailabilityPrice
Product Information
- Catalog #PopisAvailabilityPrice