Orders LinkContactUsLinkComponent
Pycnometer heads with/without bottle
Pycnometer heads with/without bottle
02, 1000 ml, 60/37
Catalog # LENZ05660007
Supplier:   Lenz Laborglas
Pycnometer heads with/without bottle
Catalog # LENZ05660007
Supplier:  Lenz Laborglas
Pycnometer heads with/without bottle
Pycnometer heads with/without bottle
02, 1000 ml, 60/37
Price
3 030,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Set, made of DURAN® borosilicate glass, consists of wide neck bottle with conical shoulder and NS joint, and pycnometer head.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for density determinations of bitumen-like and mineral materials.
Documents
Specifications
  • NZ:
    60/37
  • Objem (nom.):
    1000 ml
  • Sekvence:
    02
Accessories
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • LENZ07690070
    Bottle for pycnometer NS60/46, 1000 ml
    Log in for availability
    1 * 1 KS - 1 870,00 Kč