Orders LinkContactUsLinkComponent
Adapters, delivery, angled
Adapters, delivery, angled
14/23 NS, 65 mm
Katalogové číslo 271-2401
Dodavatel:   Lenz Laborglas
Adapters, delivery, angled
Katalogové číslo 271-2401
Dodavatel:  Lenz Laborglas
Adapters, delivery, angled
Adapters, delivery, angled
14/23 NS, 65 mm
Přejít na:
Price
612,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint socket and delivery tube at an angle of 105°. Available with long and short delivery tubes.
Documents
Specifikace
  • Velikost objímky:
    14/23 NS
  • Délka zkumavky:
    65 mm