Orders LinkContactUsLinkComponent
Multiple adapters
14/23 NS, 14/23 NS
Catalog # LENZ05015011
Supplier:   Lenz Laborglas
Multiple adapters
Catalog # LENZ05015011
Supplier:  Lenz Laborglas
Multiple adapters
Multiple adapters
14/23 NS, 14/23 NS
Price
1 800,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN® Borosilicate glass 3.3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 3 NS sockets and 1 NS cone, with two parallel necks and one angled neck (45°).
Specifications
  • Rozměry zábrusu:
    14/23 NS
  • Velikost objímky:
    14/23 NS