Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
1000 ml, 29/32 NS
Katalogové číslo 527-1510
Dodavatel:   Lenz Laborglas
Separating funnels, Gilson
Katalogové číslo 527-1510
Dodavatel:  Lenz Laborglas
Separating funnels, Gilson
Separating funnels, Gilson
1000 ml, 29/32 NS
Přejít na:
Price
4 840,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Documents
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    1000 ml