Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
100 ml, 19/26 NS
Katalogové číslo 527-1507
Dodavatel:   Lenz Laborglas
Separating funnels, Gilson
Katalogové číslo 527-1507
Dodavatel:  Lenz Laborglas
Separating funnels, Gilson
Separating funnels, Gilson
100 ml, 19/26 NS
Přejít na:
Price
2 860,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Documents
Specifikace
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    100 ml