Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
50 ml, 19/26 NS, -
Katalogové číslo 527-1321
Dodavatel:   Lenz Laborglas
Separating funnels, Squibb
Katalogové číslo 527-1321
Dodavatel:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
50 ml, 19/26 NS, -
Přejít na:
Price
1 810,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.
Documents
Specifikace
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    50 ml
  • Dělení:
    -