Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
500 ml, 29/32 NS, 10 ml
Catalog # 527-1993
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Catalog # 527-1993
Supplier:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
500 ml, 29/32 NS, 10 ml
Přejít na:
Price
2 250,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.
Documents
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    500 ml
  • Dělení:
    10 ml