Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS, -
Catalog # 527-1312
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Catalog # 527-1312
Supplier:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS, -
Price
1 480,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.
Specifications
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    100 ml
  • Dělení:
    -