Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
250 ml, 29/32 NS, -
Catalog # 527-1133
Supplier:   Lenz Laborglas
Conical separating funnels
Catalog # 527-1133
Supplier:  Lenz Laborglas
Conical separating funnels
Conical separating funnels
250 ml, 29/32 NS, -
Price
1 820,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
Specifications
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    250 ml
  • Dělení:
    -