Orders LinkContactUsLinkComponent
Rotary evaporating flasks
Rotary evaporating flasks
10, 2000 ml, 29/32 NS
Katalogové číslo 201-0328
Dodavatel:   Lenz Laborglas
Rotary evaporating flasks
Katalogové číslo 201-0328
Dodavatel:  Lenz Laborglas
Rotary evaporating flasks
Rotary evaporating flasks
10, 2000 ml, 29/32 NS
Přejít na:
Price
2 870,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
Documents
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    2000 ml
  • Sekvence:
    10