Orders LinkContactUsLinkComponent
Rotary evaporating flasks
Rotary evaporating flasks
08, 500 ml, 29/32 NS
Catalog # 201-0326
Supplier:   Lenz Laborglas
Rotary evaporating flasks
Catalog # 201-0326
Supplier:  Lenz Laborglas
Rotary evaporating flasks
Rotary evaporating flasks
08, 500 ml, 29/32 NS
Price
1 510,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
Documents
Certificates
Search by Lot Number
Specifications
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    500 ml
  • Sekvence:
    08