Rotary evaporating flasks
06, 100 ml, 29/32 NS
201-0324
DURAN®, borosilicate glass, clear or plastic-coated.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
- Velikost objímky:29/32 NS
- Kapacita:100 ml
- Sekvence:06