Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
08, Vacuum stopcocks, one way, straight, 6,0 mm, 21,5 NS
Catalog # 189-2103
Supplier:   Lenz Laborglas
Vacuum stopcocks
Catalog # 189-2103
Supplier:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
08, Vacuum stopcocks, one way, straight, 6,0 mm, 21,5 NS
Price
2 230,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifications
  • Ø otvoru:
    6,0 mm
  • Popis:
    Vacuum stopcocks, one way, straight
  • Sekvence:
    08
  • Kohout:
    21,5 NS