Stopcocks, three way
6,0 mm, 29,2 NS
Katalogové číslo 227-3235
Dodavatel: Lenz Laborglas
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys with T-bore
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
DIN 12554
Specifikace
- Ø otvoru:6,0 mm
- Kohout:29,2 NS