Stopcocks, three way
4,0 mm, 18,8 NS
Katalogové číslo 201-1036
Dodavatel: Lenz Laborglas
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys with T-bore
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
DIN 12554
Documents
BSE TSE Glass Columns
Certifikáty BSE/TSE
Specifikace
- Ø otvoru:4,0 mm
- Kohout:18,8 NS